Skip to Content
Merck
All Photos(1)

Documents

562378

Sigma-Aldrich

Benzylchlorodimethylsilane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2Si(CH3)2Cl
CAS Number:
Molecular Weight:
184.74
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.511 (lit.)

bp

101-103 °C/18 mmHg (lit.)

density

1.018 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(Cl)Cc1ccccc1

InChI

1S/C9H13ClSi/c1-11(2,10)8-9-6-4-3-5-7-9/h3-7H,8H2,1-2H3

InChI key

ABHNFDUSOVXXOA-UHFFFAOYSA-N

Application

Benzylchlorodimethylsilane can be used to prepare:
  • A ferrocene based mesoporous catalyst, applicable in the oxidation of styrene.
  • Silylbissulfide named 3-(benzyldimethylsilyl)-1,3-bis(phenylthio)-1-propene.
  • Silyl enyne by reacting with 6-hept-1-yne and nBu-Li, for further use in the intramolecular cross-metathesis to yield silicon substituted diene.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syntheses of 2-silicon-substituted 1, 3-dienes
Choudhury PP, et al.
Journal of Organometallic Chemistry, 754, 88-93 (2014)
Diversity oriented synthesis of conjugate dienes and alkenylcyclopropanes utilizing silyl group-substituted titanium carbene complexes as bimetallic synthetic reagents
Takeda T, et al.
Tetrahedron, 70(35), 5776-5786 (2014)
Ana C Gomes et al.
Dalton transactions (Cambridge, England : 2003), 42(40), 14612-14620 (2013-08-30)
The surface silanol groups in crystal-like mesoporous phenylene-silica have been derivatized with trimethylsilyl, benzyldimethylsilyl and dimethylsilyl(ferrocene) groups by performing a post-synthetic grafting reaction with the corresponding chlorosilane precursors. The success of the grafting procedure was demonstrated by transmission FT-IR spectroscopy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service