Skip to Content
Merck
All Photos(1)

Key Documents

510629

Sigma-Aldrich

Bis(ethylcyclopentadienyl)chromium(II)

Synonym(s):

1,1′-Diethylchromocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H18Cr
CAS Number:
Molecular Weight:
238.29
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

reaction suitability

core: chromium
reagent type: catalyst

density

1.15 g/mL at 25 °C (lit.)

SMILES string

[Cr].CC[C]1[CH][CH][CH][CH]1.CC[C]2[CH][CH][CH][CH]2

InChI

1S/2C7H9.Cr/c2*1-2-7-5-3-4-6-7;/h2*3-6H,2H2,1H3;

InChI key

OZUJAILVLUFQME-UHFFFAOYSA-N

Application

Catalyst for polymerization of ethylene

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Pyr. Liq. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service