Skip to Content
Merck
All Photos(1)

Key Documents

487260

Sigma-Aldrich

2-Chlorobenzyl isocyanate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
RM 486.00

RM 486.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
RM 486.00

About This Item

Linear Formula:
ClC6H4CH2NCO
CAS Number:
Molecular Weight:
167.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

RM 486.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.547 (lit.)

bp

238 °C (lit.)

density

1.149 g/mL at 25 °C (lit.)

functional group

amine
chloro
isocyanate

storage temp.

2-8°C

SMILES string

Clc1ccccc1CN=C=O

InChI

1S/C8H6ClNO/c9-8-4-2-1-3-7(8)5-10-6-11/h1-4H,5H2

InChI key

IBRWLEYNCGRJLX-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service