Skip to Content
Merck
All Photos(1)

Key Documents

434418

Sigma-Aldrich

Phenol-d6

≥99 atom % D

Synonym(s):

Hexadeuterophenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5OD
CAS Number:
Molecular Weight:
100.15
Beilstein:
2255089
EC Number:
MDL number:
UNSPSC Code:
12352200

vapor density

3.24 (vs air)

vapor pressure

0.36 mmHg ( 20 °C)

isotopic purity

≥99 atom % D

autoignition temp.

1319 °F

expl. lim.

8.6 %

bp

182 °C (lit.)

mp

40-42 °C (lit.)

mass shift

M+6

SMILES string

[2H]Oc1c([2H])c([2H])c([2H])c([2H])c1[2H]

InChI

1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H/i1D,2D,3D,4D,5D/hD

InChI key

ISWSIDIOOBJBQZ-QNKSCLMFSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in environmental applications.

Packaging

Packaged in prescored ampules

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service