Skip to Content
Merck
All Photos(1)

Documents

310018

Sigma-Aldrich

Pentamethylenebis(magnesium bromide) solution

0.5 M in THF

Synonym(s):

Pentane-1,5-di(magnesium bromide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrMg(CH2)5MgBr
CAS Number:
Molecular Weight:
278.55
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

density

0.999 g/mL at 25 °C

SMILES string

Br[Mg]CCCCC[Mg]Br

InChI

1S/C5H10.2BrH.2Mg/c1-3-5-4-2;;;;/h1-5H2;2*1H;;/q;;;2*+1/p-2

InChI key

OUEGHZYBWJTUAD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Other Notes

Storage below 25°C may cause formation of crystalline magnesium salts. Moving container to a warm location and occasional gentle swirling should redissolve the solid

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 2

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cameron S Cowan et al.
Cell, 182(6), 1623-1640 (2020-09-19)
Human organoids recapitulating the cell-type diversity and function of their target organ are valuable for basic and translational research. We developed light-sensitive human retinal organoids with multiple nuclear and synaptic layers and functional synapses. We sequenced the RNA of 285,441

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service