Skip to Content
Merck
All Photos(1)

Documents

305588

Sigma-Aldrich

Butylphosphonic dichloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3P(O)Cl2
CAS Number:
Molecular Weight:
174.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.466 (lit.)

bp

96-97 °C/15 mmHg (lit.)

density

1.241 g/mL at 25 °C (lit.)

SMILES string

CCCCP(Cl)(Cl)=O

InChI

1S/C4H9Cl2OP/c1-2-3-4-8(5,6)7/h2-4H2,1H3

InChI key

YITVQRWHXAYYIX-UHFFFAOYSA-N

General description

Butylphosphonic dichloride is a phosphorous halide.

Application

Butylphosphonic dichloride was used in preparation of aryl-phosphonic ester, required for synthesis of organophosphorous polymers.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organophosphorus polymers Communication 1. polymerization of cyclic alkyl-and aryl-phosphonic esters.
Korshak VV, et al.
Russian Chemical Bulletin, 6(5), 641-646 (1957)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service