Skip to Content
Merck
All Photos(1)

Key Documents

256978

Sigma-Aldrich

Ethylmagnesium bromide solution

3.0 M in diethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2MgBr
CAS Number:
Molecular Weight:
133.27
Beilstein:
3587203
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

reaction suitability

reaction type: Grignard Reaction

concentration

3.0 M in diethyl ether

density

1.02 g/mL at 25 °C

SMILES string

CC[Mg]Br

InChI

1S/C2H5.BrH.Mg/c1-2;;/h1H2,2H3;1H;/q;;+1/p-1

InChI key

TWTWFMUQSOFTRN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Recommended products

Angled septum-inlet adapter Z118206 or straight septum-inlet adapter, 6mm I.D. inlet, Z118141 or straight septum-inlet adapter, 13mm I.D. inlet, Z118192 is recommended.

septum inlet adapter

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Nishiyama et al.
Chemical & pharmaceutical bulletin, 48(12), 1999-2002 (2001-01-06)
The direct alpha-bromination of various ketones using trifluoromethanesulfonic anhydride and Grignard reagent or magnesium bromide in ether gave the corresponding alpha-bromo ketones in moderate to good yields under mild reaction conditions.
F Turon et al.
Lipids, 37(8), 817-821 (2002-10-10)
An analytical procedure was developed for regiodistribution analysis of TAG using alpha-MAG prepared by an ethyl magnesium bromide deacylation. In the present communication, the deacylation procedure is shown to lead to representative alpha-MAG, allowing the composition of the native TAG
R G Xie et al.
Steroids, 55(11), 488-490 (1990-11-01)
The reaction of ethyl magnesium bromide and 17 alpha-ethynylestradiol with formaldehyde in the presence of triethyl phosphate or hexamethylphosphoramide gave the 2- and 4-formyl-17 alpha-ethynylestradiol in high yield. Treatment of the formyl derivatives with an alkaline solution of hydrogen peroxide
Yuichi Kobayashi et al.
Organic letters, 7(2), 183-186 (2005-01-14)
[Reaction: see text] Low regioselectivity of RMgBr (R = aryl, alkenyl) in the CuCN-catalyzed reaction with 4-cyclopentene-1,3-diol monoacetate is improved by addition of LiCl or MgCl2 to a similar extent as previously obtained with RMgCl (>90:10). The limitation encountered in
Janne E Tønder et al.
Organic & biomolecular chemistry, 2(10), 1447-1455 (2004-05-12)
Copper mediated allylic substitutions and conjugate additions to geranyl, cinnamyl and allylic indole compounds have been investigated with the aim of finding a method for the creation of the all-carbon quaternary centre present in the natural product lyngbyatoxin A. Reaction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service