All Photos(1)
About This Item
Linear Formula:
Cl2C6H3CONH2
CAS Number:
Molecular Weight:
190.03
Beilstein:
1869103
EC Number:
MDL number:
UNSPSC Code:
12352103
Assay
97%
mp
196-199 °C (lit.)
SMILES string
NC(=O)c1c(Cl)cccc1Cl
InChI
1S/C7H5Cl2NO/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,(H2,10,11)
InChI key
JHSPCUHPSIUQRB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service