Merck
Apoptotic human neutrophil peptide-1 anti-tumor activity revealed by cellular biomechanics.
Biochimica et biophysica acta (2014-12-03)
Diana Gaspar, João M Freire, Teresa R Pacheco, João T Barata, Miguel A R B Castanho
RESUMEN
Cancer remains a major cause of morbidity and mortality worldwide. Although progress has been made regarding chemotherapeutic agents, new therapies that combine increased selectivity and efficacy with low resistance are still needed. In the search for new anticancer agents, therapies based on biologically active peptides, in particular, antimicrobial peptides (AMPs), have attracted attention for their decreased resistance development and low cytotoxicity. Many AMPs have proved to be tumoricidal agents against human cancer cells, but their mode of action is still controversial. The existence of common properties shared by the membranes of bacteria and tumor cells points to similar lipid-targeting mechanisms in both cases. On the other hand, anticancer peptides (ACPs) also induce apoptosis and inhibit angiogenesis. Human neutrophil peptide-1 (HNP-1) is an endogenous AMP that has been implicated in different cellular phenomena such as tumor proliferation. The presence of HNP-1 in the serum/plasma of oncologic patients turns this peptide into a potential tumor biomarker. The present work reveals the different effects of HNP-1 on the biophysical and nanomechanical properties of solid and hematological tumor cells. Studies on cellular morphology, cellular stiffness, and membrane ultrastructure and charge using atomic force microscopy (AFM) and zeta potential measurements show a preferential binding of HNP-1 to solid tumor cells from human prostate adenocarcinoma when compared to human leukemia cells. AFM also reveals induction of apoptosis with cellular membrane defects at very low peptide concentrations. Understanding ACPs mode(s) of action will certainly open innovative pathways for drug development in cancer treatment.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Diethyl azodicarboxylate solution, 40 wt. % in toluene
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Streptomycin Ready Made Solution, 100 mg/mL in water
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad