Merck
Neuron-Glia Interactions in Tuberous Sclerosis Complex Affect the Synaptic Balance in 2D and Organoid Cultures.
Cells (2021-01-16)
Stephanie Dooves, Arianne J H van Velthoven, Linda G Suciati, Vivi M Heine
RESUMEN
Tuberous sclerosis complex (TSC) is a genetic disease affecting the brain. Neurological symptoms like epilepsy and neurodevelopmental issues cause a significant burden on patients. Both neurons and glial cells are affected by TSC mutations. Previous studies have shown changes in the excitation/inhibition balance (E/I balance) in TSC. Astrocytes are known to be important for neuronal development, and astrocytic dysfunction can cause changes in the E/I balance. We hypothesized that astrocytes affect the synaptic balance in TSC. TSC patient-derived stem cells were differentiated into astrocytes, which showed increased proliferation compared to control astrocytes. RNA sequencing revealed changes in gene expression, which were related to epidermal growth factor (EGF) signaling and enriched for genes that coded for secreted or transmembrane proteins. Control neurons were cultured in astrocyte-conditioned medium (ACM) of TSC and control astrocytes. After culture in TSC ACM, neurons showed an altered synaptic balance, with an increase in the percentage of VGAT+ synapses. These findings were confirmed in organoids, presenting a spontaneous 3D organization of neurons and glial cells. To conclude, this study shows that TSC astrocytes are affected and secrete factors that alter the synaptic balance. As an altered E/I balance may underlie many of the neurological TSC symptoms, astrocytes may provide new therapeutic targets.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Laminina from Engelbreth-Holm-Swarm murine sarcoma basement membrane, 1-2 mg/mL in Tris-buffered saline, 0.2 μm filtered, BioReagent, suitable for cell culture
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
200 μg
Fecha estimada de envío 23 de marzo de 2025
Está disponible un anticuerpo recombinante y sin conservantes para su diana. Pruebe ZRB1226
USD 660.00
Sigma-Aldrich
Insulina solution human, sterile-filtered, BioXtra, suitable for cell culture
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
200 μg
Fecha estimada de envío 23 de marzo de 2025
Está disponible un anticuerpo recombinante y sin conservantes para su diana. Pruebe ZRB1226
USD 660.00
Sigma-Aldrich
Ácido retinoico, ≥98% (HPLC), powder
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
200 μg
Fecha estimada de envío 23 de marzo de 2025
Está disponible un anticuerpo recombinante y sin conservantes para su diana. Pruebe ZRB1226
USD 660.00
Sigma-Aldrich
Penicilina- Estreptomicina, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
200 μg
Fecha estimada de envío 23 de marzo de 2025
Está disponible un anticuerpo recombinante y sin conservantes para su diana. Pruebe ZRB1226
USD 660.00
Sigma-Aldrich
DAPI, for nucleic acid staining
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
200 μg
Fecha estimada de envío 23 de marzo de 2025
Está disponible un anticuerpo recombinante y sin conservantes para su diana. Pruebe ZRB1226
USD 660.00
Sigma-Aldrich
Poly-L-ornithine hydrobromide, mol wt 30,000-70,000
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
200 μg
Fecha estimada de envío 23 de marzo de 2025
Está disponible un anticuerpo recombinante y sin conservantes para su diana. Pruebe ZRB1226
USD 660.00
Sigma-Aldrich
Monofosfato de N6,2′-O-dibutiriladenosina 3′,5′-cíclico sodium salt, ≥97% (HPLC), powder
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
200 μg
Fecha estimada de envío 23 de marzo de 2025
Está disponible un anticuerpo recombinante y sin conservantes para su diana. Pruebe ZRB1226
USD 660.00
Sigma-Aldrich
5-Bromo-2′-desoxiuridina, ≥99% (HPLC)
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
200 μg
Fecha estimada de envío 23 de marzo de 2025
Está disponible un anticuerpo recombinante y sin conservantes para su diana. Pruebe ZRB1226
USD 660.00
Sigma-Aldrich
3,3′,5-Triiodo-L-thyronine sodium salt, powder, BioReagent, suitable for cell culture
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
200 μg
Fecha estimada de envío 23 de marzo de 2025
Está disponible un anticuerpo recombinante y sin conservantes para su diana. Pruebe ZRB1226
USD 660.00
Sigma-Aldrich
Ácido L-ascórbico, suitable for cell culture, suitable for plant cell culture, ≥98%
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
200 μg
Fecha estimada de envío 23 de marzo de 2025
Está disponible un anticuerpo recombinante y sin conservantes para su diana. Pruebe ZRB1226
USD 660.00
Sigma-Aldrich
Ácido valproico sodium salt, 98%
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
200 μg
Fecha estimada de envío 23 de marzo de 2025
Está disponible un anticuerpo recombinante y sin conservantes para su diana. Pruebe ZRB1226
USD 660.00
Sigma-Aldrich
Triton X-100, laboratory grade
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
200 μg
Fecha estimada de envío 23 de marzo de 2025
Está disponible un anticuerpo recombinante y sin conservantes para su diana. Pruebe ZRB1226
USD 660.00
Sigma-Aldrich
Heparina sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
200 μg
Fecha estimada de envío 23 de marzo de 2025
Está disponible un anticuerpo recombinante y sin conservantes para su diana. Pruebe ZRB1226
USD 660.00
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización
Número de referencia del producto (SKU)Tamaño de envaseDisponibilidadPrecioCantidad
200 μg
Fecha estimada de envío 23 de marzo de 2025
Está disponible un anticuerpo recombinante y sin conservantes para su diana. Pruebe ZRB1226
USD 660.00