Documentos clave
SU860131
Supelco
Screw Cap with Hole
thread for 8-425
About This Item
UNSPSC Code:
12352200
NACRES:
NB.11
material
black polypropylene (with 5.5 mm center hole)
feature
autoclavable
packaging
pkg of 1000 ea
parameter
135 °C temp. limit
fitting
thread for 8-425
compatibility
for use with HITACHI
General description
These hole caps do not come with septa.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)