Documentos
ES5334
GreenSep PFP SFC 5 Micron HPLC Column
5 μm particle size, L × I.D. 15 cm × 20 mm
About This Item
UNSPSC Code:
12352200
NACRES:
SB.52
material
stainless steel column
product line
GreenSep
manufacturer/tradename
ES Industries 138291-GS-PFP
packaging
1 ea of
technique(s)
HPLC: suitable
L × I.D.
15 cm × 20 mm
matrix
fully porous particle
matrix active group
PFP (pentafluorophenyl) phase
particle size
5 μm
pore size
120 Å pore size
operating pH
2-8
separation technique
supercritical fluid (SFC)
General description
Specifically designed for the separation of geometrical isomers as well as diastereomers. It is the column of choice in separating compounds that contain aromatic groups, polarizable electrons and conjugate systems. In addition, GreenSep PFP is useful for the separation of halogenated compounds. In many cases GreenSep PFP provides orthogonal separations when compared to GreenSep Nitro. GreenSep PFP is unsurpassed for its separation capabilities and is available in analytical columns, preparative columns and bulk media.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
GreenSep is a trademark of ES Industries
Choose from one of the most recent versions:
Certificados de análisis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Atención al cliente