Documentos
20592
Glass Tubing Cutter Replacement Wheels
pkg of, pkg of 5 ea
About This Item
UNSPSC Code:
41122406
packaging
pkg of 5 ea
pkg of
compatibility
for use with 20591
Choose from one of the most recent versions:
Certificados de análisis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Atención al cliente