Documentos clave
Z743562
Write protect tape
clear, L × W 1.6 in. × 0.63 in. , perforated to fit 0.5 mL and 1.5 mL tubes
Seleccione un Tamaño
1 EA
MXP 777.00
MXP 777.00
Fecha estimada de envío22 de mayo de 2025
Seleccione un Tamaño
Cambiar Vistas
1 EA
MXP 777.00
About This Item
Código UNSPSC:
41122700
NACRES:
NB.17
MXP 777.00
Fecha estimada de envío22 de mayo de 2025
envase
pack of 1 ea
fabricante / nombre comercial
Diversified Biotech WRIT-2000
L × An
1.6 in. × 0.63 in. , perforated to fit 0.5 mL and 1.5 mL tubes
color
clear
Descripción general
A Clear Adhesive Overwrap to Protect Writing
Resistant - Defend writing against organic solvents and caustic agents
Easy Application - Comes in a handy dispenser and is perforated to fit 0.5ml and 1.5ml tubes.
Multi-Surface - Adhere to all plastics, glass and metals
Temperature Range - -196 C to 150 C
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)