Documentos clave
Z688126
E-Vac polypropylene bottle
with standard lid, capacity 4,000 mL
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material
polypropylene bottle
round bottle
joint
joint threaded
feature
without cap
packaging
pack of 1 ea
manufacturer/tradename
Cole Parmer 1305040
capacity
4,000 mL
General description
The E-Vac bottles can be autoclaved for 20 minutes at 121°C to prevent laboratory contamination. Base units are illuminated to allow visual inspection of waste levels in the bottle. Dual hydrophobic filters prevent liquids from reaching the pump housing and contaminating the system.
E-Vac is available with a 4-L polypropylene bottle or a 3-L safety glass bottle. Polypropylene and glass bottles offer lids equipped with quick-release tubing connectors for safe and convenient tubing connections; and level detection sensor that automatically shuts the pump off when the bottle is full. Polypropylene bottle also available with lid fitted with standard barbed tube fittings.
For use with
Referencia del producto
Descripción
Precios
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)