Documentos clave
Z614033
Vivaspin 500 centrifugal concentrators
MWCO 10 kDa
About This Item
Código UNSPSC:
41104916
NACRES:
NB.24
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Materiales
polycarbonate housing
polyethersulfone membrane
descripción
PES membrane
esterilidad
non-sterile
Características
holdup volume 5 μL
envase
package of 100 ea
fabricante / nombre comercial
Sartorius VS0102
Parámetros
15000 × g max. centrifugal force
volumen de bloqueo
5 μL
área de la membrana
0.5 cm2
tamaño tubo
~2.2 mL
volumen de trabajo
500 μL
tamaño de poro
10 kDa MWCO
Descripción general
  • Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs.
  • Highest flow rates and recoveries are achieved due to their particular vertical membrane design.
  • Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin.
Note: Vivaspin 500 μL centrifugal filter units offer a simple one step procedure for sample preparation. They can effectively be used in either swing out or fixed angle rotors.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)