Documentos
Z369012
Sigma-Aldrich
Finntip® Micro-tips 10 μL capacity
non-sterile, pack of 960 ea (10 racks of 96 tips)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
material
pink
sterility
non-sterile
feature
barrier: no
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Finn 9400300
L
3.2 cm
maximum volume
10 μL
volume range
0.1-10 μL
Compatibility
Tips for use specifically with micro-volume Finnpipettes
Legal Information
Finntip is a registered trademark of Thermo Fisher Scientific or its subsidiaries