Z359769
Peptide Antigens: A Practical Approach
About This Item
ISBN-10:
0-19-963451-3
ISBN-13:
978-0-19-963451-4
Código UNSPSC:
55101509
info de publicaciones
G.B. Wisdom, ed., IRL Press at Oxford University Press, Oxford, England, 1994, 252 pp., soft cover
fabricante / nombre comercial
Oxford University Press
Características y beneficios
This volume provides practical guidance to the major techniques used in the exploitation of peptide antigens and anti-peptide antibodies. The chapters give detailed protocols for the prediction of epitopes, peptide synthesis, the preparation of peptide immunogens, immunoaffinity chromatography, immunoassays, and the mapping of epitopes using both synthetic peptides and phage display systems.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)