Documentos clave
Z317322
Eppendorf® racks for microcentrifuge tubes
For 24 × 0.5 ml tubes
About This Item
UNSPSC Code:
41122804
manufacturer/tradename
Eppendorf® 0030 123.107
availability
not available in USA and Canada
General description
  • 24-place capacity
  • Unique swivel connector to interlock multiple racks for front-to-back and side-to-side configurations
  • Non-slip stand
  • Numbered positions
  • UV-resistant
  • Autoclavable
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)