Documentos clave
T0566
Dounce tissue grinder tube
working volume 7 mL
Seleccione un Tamaño
1 EA
MXP 2,008.00
MXP 2,008.00
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
MXP 2,008.00
About This Item
Código UNSPSC:
41121800
NACRES:
NB.22
MXP 2,008.00
Check Cart for Availability
Materiales
glass
envase
pack of 1 ea
volumen de trabajo
7 mL
Descripción general
Designed primarily for cellular work where the nucleus remains intact after homogenization.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Questions
  1. I would like to ask you regarding the product https://www.sigmaaldrich.com/GR/en/product/sigma/t0566 Which pestle is suitable for the particular grinder?
    1 answer
    1. There are two pestle options for the 7 mL tissue grinder.
      See below for a link to each product.
      P1235, Small Clearance Pestle
      https://www.sigmaaldrich.com/product/sigma/p1235
      P0610, Large Clearance Pestle
      https://www.sigmaaldrich.com/product/sigma/p0610