Documentos clave
HS120113
Mega Rack® tube rack
double, for 10-13 mm tubes, blue
Seleccione un Tamaño
1 EACH
MXP 1,526.00
MXP 1,526.00
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EACH
MXP 1,526.00
About This Item
Código UNSPSC:
41122804
NACRES:
NB.12
MXP 1,526.00
Check Cart for Availability
Materiales
polypropylene
Características
Holds 432 x 5-7 mL tubes
envase
pack of 1 ea
fabricante / nombre comercial
Heathrow Scientific HS120113
tamaño
673x235x90 mm
color
blue
Descripción general
Mega Rack High-capacity storage rack for sample and blood collection tubes.
  • Fully-assembled, polypropylene Mega Tube Racks are great for high throughput labs requiring large sample storage.
  • Sturdy and light weight material for easy transport
  • Alphanumeric identification for quick location and retrieval of tubes
  • Place different test groupings into one rack and eliminate need for multiple tube racks
  • Autoclavable
Información legal
Mega Rack is a registered trademark of Heathrow Scientific
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)