Documentos
GE18117263
Fingertight Connector 1/16" Male
ÄKTA®express, Cytiva 18117263
About This Item
UNSPSC Code:
41115705
NACRES:
NB.43
material
black PEEK
feature
Connections: Fingertight 1/16"
Wetted Material: Polyetheretherketone (PEEK)
ÄKTA®express
manufacturer/tradename
Cytiva 18117263
compatibility
For use with: Tubing O.D. 1/16"
Categorías relacionadas
Legal Information
ÄKTA is a registered trademark of Cytiva