Documentos clave
GE10600030
Amersham Hybond® SEQ Western blotting membranes, PVDF
pore size 0.2 μm, pkg of 1 ea
About This Item
UNSPSC Code:
41105339
NACRES:
NB.22
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 10600030
pore size
0.2 μm pore size
General description
PVDF protein membrane, specifically designed for protein sequencing applications and solid phase amino acid analysis.
Features and Benefits
  • High protein binding capacity for easy signal detection.
  • Chemical resistance needed for N-terminal sequencing.
  • High protein retention even after harsh washing steps.
  • Efficient capture of proteins during transfer minimizes sample loss.
  • 0.2 µm pore size for higher surface area, resulting in better binding of low molecular weight proteins.
  • Available in popular precut sizes for your application.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Legal Information
Amersham is a trademark of Cytiva
Hybond is a registered trademark of Cytiva
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)