Documentos
EP5424703003
Eppendorf® Spare lid for rotor FA-45-24-11
aerosol tight
About This Item
UNSPSC Code:
41103913
NACRES:
NB.82
feature
aerosol tight
manufacturer/tradename
5424703003
Eppendorf® 5424703003
Eppendorf®
technique(s)
centrifugation: suitable
compatibility
for use with 5424 R
General description
Designed for use with Eppendorf rotor FA–45-24-11 this lid provides an aerosol tight seal.
Features and Benefits
  • Spare Rotor Lid
  • Aerosol tight
  • For use with liquid handling applications
Legal Information
Eppendorf is a registered trademark of Eppendorf AG