Documentos clave
EP4924000304
Eppendorf® Reference 2 G, BASIC, single-channel, variable
volume 0.5-5 mL, purple
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
El producto EP4924000304 no se vende actualmente en su país Póngase en contacto con el Servicio técnico
Características
single-channel
envase
pack of 1 ea
pkg of 1 ea
fabricante / nombre comercial
Eppendorf® 4924000304
volumen
0.5-5 mL
color
purple
Descripción general
The name, Reference, stands for extraordinary precision and accuracy, a long service lifes and an ergonomic design. The new Reference 2 boasts these proven premium characteristics with its state-of-the-art technolgy, making it a reliable partner for you and your work. The time saving single-button operation is unrivalled in the new multichannel versions.
Información legal
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Artículos
Analysis of heavy metals and metals in cannabis and hemp containing personal care products and cosmetics include sample preparation by grinding, preparation of calibration solutions for a standard addition approach, a microwave digestion protocol and subsequent ICP-MS analysis.
Protocolos
A complete SEC-UV workflow for quantifying monoclonal antibody fragments, monomers, and aggregates, including procedure for a system suitability test and heat stressing of antibodies.