Documentos clave
EP0030742060
Eppendorf® Cell Imaging Slides
Tissue Culture (TC)-treated surface, pack of 16 plates (individually wrapped), wells 4
About This Item
Código UNSPSC:
41122600
Materiales
colorless
soda-lime glass
esterilidad
sterile
envase
pack of 16 plates (individually wrapped)
fabricante / nombre comercial
Eppendorf® 0030742060
An × P × Al
× 76.0 mm × 12.4 mm
pocillos
4
volumen de trabajo
0.7-1.0 mL
tipo de unión
Tissue Culture (TC)-treated surface
Descripción general
The easy-to-open, securely closing, Flex Tubes offers comfort and dependability for sample preparation, centrifugation, and storage. One vessel for all applications in molecular biology, chemistry, and biochemistry.
Safer than ever: Flex Tubes can be centrifuged up to 30,000 × g without any problems!
Best material!
Eppendorf consumables incl. the Eppendorf Tubes 3810X are made exclusively from high quality, ultra pure polypropylene, which complies with FDA guidelines 21 CFR § 177.1520 »Olefin Polymers«, and 21 CFR § 178.2010 »Antioxidants and Stabilizers for Polymers«. Extensive quality checks are in place prior to and during production. Furthermore, the final product is examined for functionality. The functionality tests for tubes include lid closure security in a boiling water bath, centrifugation stability and vapor tightness. These quality controls and tests ensure the sustained high product quality across the entire production phase – lot by lot. Please see APPLICATION NOTE No.056 for details: The best material for original Eppendorf Tubes and Plates: Properties and chemical resistance of polypropyleneFlex Tubes
Highest sample integrity!
Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio–assay results or measurements, e.g. nucleic acid quantification by photometric measurements at 260 nm.
Características y beneficios
Tube lid is easy to open and close for ergonomic operation
Polypropylene provides high resistance to chemicals, mechanical stress, and temperature extremes
g-Safe: Exceptional centrifugation stability up to 30,000 × g prevents sample loss due to tube breakage and provides extra safety when working with hazardous samples
Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results
Graduation for simple verification of the pipetted volume
Frosted lid and surface on the side for easy labeling
Guaranteed functionality from -86 deg. C to 100 deg. C
Autoclavable when open (121 deg.C, 20 min)
Available in Eppendorf Quality and lot-certified PCR clean purity grade
Información legal
Eppendorf is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)