EP0030075099
Eppendorf® epT.I.P.S.® G Racks, Biopur®
size (50-1250 μL), L (76 mm), green, pack of 480 ea (5 trays of 96 tips)
About This Item
Materiales
green (Tray)
esterilidad
sterile
Características
DNase free
envase
pack of 480 ea (5 trays of 96 tips)
fabricante / nombre comercial
Eppendorf® 0030075099
Longitud
(76 mm)
volumen máximo
1.25 mL
tamaño
(50-1250 μL)
volumen
50—1250 μL
color
green
Descripción general
- Eppendorf® epT.I.P.S.® G Racks, Biopur® offer the highest biological purity grade, Guaranteed PCR clean, sterile, pyrogen-, ATP- and DNA-free.
- For use in medical, pharmaceutical and food industry, molecular biology and cell technology.
- Each production lot inspected continuously by independent laboratory.
- Packed in racks with 96 tips.
Información legal
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)