CLS7605
Corning® Transtar® liquid handling system
adjustable volume pipettor
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
Productos recomendados
esterilidad
non-sterile
fabricante / nombre comercial
Corning 7605
volumen
200 μL
Descripción general
The Transtar is a portable, autoclavable liquid handling system designed for use with 96 well plates, A sterile 96 tip disposable cartridge, which loads into the Transtar system, enables liquids to be aspirated, transferred and dispensed over a volume range of 25 - 200 μL in 5 μL increments, Transtar accuracy us rated at 5% at all volume levels.
Información legal
Corning is a registered trademark of Corning, Inc.
TRANSTAR-96 is a registered trademark of Corning, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)