Merck
C5916
Sigma-Aldrich
Culture tubes
size 25 mm × 150 mm, borosilicate glass
Sinónimos:
culture tube, test tube, test tubes
About This Item
Código UNSPSC:
41100000
Materiales
borosilicate glass
cap (Test tube cap)
esterilidad
non-sterile
envase
case of 72 tubes
tamaño
25 mm × 150 mm
Características y beneficios
  • Kimble® culture tubes are manufactured from low extractable, chemical-resistant borosilicate glass.
  • These reusable tubes feature a fire-polished top rim.
  • Uniform wall thickness ensures maximum heat transfer.
  • The marking spot is printed in durable white ceramic enamel.
Envase
Packaged in a sturdy cardboard box with internal grid to minimize damage during shipping and handling.
Información legal
KIMBLE is a registered trademark of DWK Life Sciences
Opcional
Referencia del producto
Descripción
Precios
sustituido por
Referencia del producto
Descripción
Precios
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)