Saltar al contenido
Merck

79260

Supelco

(S)-3-(4-Chloro-3-fluorophenoxy)-2-hydroxy-2-methyl-N-(4-nitro-3-trifluoromethylphenyl)propionamide

analytical standard

Sinónimos:

S-6

Iniciar sesiónpara Ver la Fijación de precios por contrato y de la organización


About This Item

Fórmula empírica (notación de Hill):
C17H13ClF4N2O5
Número de CAS:
Peso molecular:
436.74
Beilstein:
9733029
Número MDL:
Código UNSPSC:
41116107
ID de la sustancia en PubChem:
NACRES:
NA.24

grado

analytical standard

Nivel de calidad

Ensayo

≥98.0% (HPLC)

pureza óptica

enantiomeric excess: ≥96.0%

caducidad

limited shelf life, expiry date on the label

composición

carbon content, 46.75%
hydrogen content, 3.00%
nitrogen content, 6.41%

técnicas

HPLC: suitable
gas chromatography (GC): suitable

aplicaciones

forensics and toxicology
pharmaceutical (small molecule)

Formato

neat

cadena SMILES

C[C@](O)(COc1ccc(Cl)c(F)c1)C(=O)Nc2ccc(c(c2)C(F)(F)F)[N+]([O-])=O

InChI

1S/C17H13ClF4N2O5/c1-16(26,8-29-10-3-4-12(18)13(19)7-10)15(25)23-9-2-5-14(24(27)28)11(6-9)17(20,21)22/h2-7,26H,8H2,1H3,(H,23,25)/t16-/m0/s1

Clave InChI

LVWMWYCDQNZFKV-INIZCTEOSA-N

Aplicación

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Envase

Bottomless glass bottle. Contents are inside inserted fused cone.

Código de clase de almacenamiento

11 - Combustible Solids

Clase de riesgo para el agua (WGK)

WGK 3

Punto de inflamabilidad (°F)

Not applicable

Punto de inflamabilidad (°C)

Not applicable


Elija entre una de las versiones más recientes:

Certificados de análisis (COA)

Lot/Batch Number

¿No ve la versión correcta?

Si necesita una versión concreta, puede buscar un certificado específico por el número de lote.

¿Ya tiene este producto?

Encuentre la documentación para los productos que ha comprado recientemente en la Biblioteca de documentos.

Visite la Librería de documentos

Yuan Zhang et al.
Biochimica et biophysica acta, 1861(2), 130-137 (2015-11-17)
Atypical angiopoietin-like 8 (ANGPTL8), also known as betatrophin, is known to regulate lipid metabolism. However, its mechanism of action remains elusive. HepG2, 3T3-L1, and NIT-1 cells were cultured in amino acid-complete MEM or histidine-free MEM to detect ANGPTL8 expression. The
Elena Sánchez-López et al.
Scientific reports, 9(1), 806-806 (2019-01-30)
Capillary electrophoresis-mass spectrometry (CE-MS) using a sheathless porous tip interface emerged as an attractive tool in metabolomics thanks to its numerous advantages. One of the main advantages compared to the classical co-axial sheath liquid interface is the increased sensitivity, while
Xiao-Yan Liu et al.
Journal of separation science, 43(23), 4247-4262 (2020-09-26)
A method of ultra-fast liquid chromatography in series with tandem mass spectrometry for the rapid and sensitive detection of 57 compounds in Spatholobi Caulis (the vine stem of Spatholobus suberectus Dunn) within 35 min was established. This assay can simultaneously determine
Jianjun Han et al.
Oncotarget, 8(67), 110965-110982 (2018-01-18)
Obesity is a significant risk factor for ovarian cancer (OC) and associated with worse outcomes for this disease. We assessed the anti-tumorigenic effects of metformin in human OC cell lines and a genetically engineered mouse model of high grade serous
José Carlos Arévalo-Lorido et al.
Vascular, 27(1), 19-26 (2018-09-13)
The role of serum uric acid in ischemic stroke is controversial. On the one hand, it has a role as neuroprotectant in acute phase, but on the other hand, it may promote atherosclerosis in carotid arteries. Our aim is to

Questions

Reviews

No rating value

Active Filters

Nuestro equipo de científicos tiene experiencia en todas las áreas de investigación: Ciencias de la vida, Ciencia de los materiales, Síntesis química, Cromatografía, Analítica y muchas otras.

Póngase en contacto con el Servicio técnico