Merck
66610
Sigma-Aldrich
N-Methyldiethanolamine
purum, ≥98.0% (GC)
Sinónimos:
2,2′-Methyliminodiethanol, N,N-Bis(2-hydroxyethyl)methylamine, MDEA
About This Item
Fórmula lineal:
CH3N(CH2CH2OH)2
Número de CAS:
Peso molecular:
119.16
Beilstein:
1734441
Número CE:
Número MDL:
Código UNSPSC:
12352100
ID de la sustancia en PubChem:
densidad de vapor
4 (vs air)
presión de vapor
0.01 mmHg ( 20 °C)
grado
purum
Ensayo
≥98.0% (GC)
temp. de autoignición
770 °F
lim. expl.
99 %, 55 °F
~56 %, 23 °F
~8.8 %
índice de refracción
n20/D 1.469 (lit.)
n20/D 1.469
bp
246-248 °C (lit.)
densidad
1.038 g/mL at 25 °C (lit.)
cadena SMILES
CN(CCO)CCO
InChI
1S/C5H13NO2/c1-6(2-4-7)3-5-8/h7-8H,2-5H2,1H3
Clave InChI
CRVGTESFCCXCTH-UHFFFAOYSA-N
Pictogramas
Exclamation mark
Palabra de señalización
Warning
Frases de peligro
Clasificaciones de peligro
Eye Irrit. 2
Código de clase de almacenamiento
10 - Combustible liquids
Clase de riesgo para el agua (WGK)
WGK 1
Punto de inflamabilidad (°F)
260.6 °F - closed cup
Punto de inflamabilidad (°C)
127 °C - closed cup
Equipo de protección personal
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Jacob D McDonald et al.
Environmental science & technology, 48(18), 10821-10828 (2014-08-29)
Carbon dioxide (CO2) absorption with aqueous amine solvents is a method of carbon capture and sequestration (CCS) from flue gases. One concern is the possible release of amine solvents and degradation products into the atmosphere, warranting evaluation of potential pulmonary
N Bord et al.
Analytical and bioanalytical chemistry, 380(2), 325-332 (2004-09-01)
Alkanolamines such as diethanolamine (DEA) and N-methyldiethanolamine (MDEA) are used in desulfurization processes in crude oil refineries. These compounds may be found in process waters following an accidental contamination. The analysis of alkanolamines in refinery process waters is very difficult
Márcio Rodrigues et al.
Journal of pharmacy & pharmaceutical sciences : a publication of the Canadian Society for Pharmaceutical Sciences, Societe canadienne des sciences pharmaceutiques, 17(3), 302-315 (2014-09-17)
Carica papaya has been traditionally used worldwide in folk medicine to treat a wide range of ailments in humans, including the management of obesity and digestive disorders. However, scientific information about its potential to interact with conventional drugs is lacking.
Camilla Montesano et al.
Talanta, 130, 382-387 (2014-08-28)
The binding specificity of a bio-inspired hexapeptide (QHWWDW) versus cocaine and four other drugs such as 3,4-methylenedioxy-N-methylamphetamine (MDMA), 3,4-methylenedioxy-N-ethylamphetamine (MDEA), phencyclidine and morphine was computationally studied and then experimentally confirmed in solid phase extraction (SPE) followed by liquid chromatography-mass spectrometry
G L Gresham et al.
Journal of mass spectrometry : JMS, 35(12), 1460-1469 (2001-02-17)
The nitrogen blister agents HN-2 (bis(2-chloroethyl)methylamine) and HN-3 (tris(2-chloroethyl)amine) were directly analyzed on the surface of soil samples using ion trap secondary ion mass spectrometry (SIMS). In the presence of water, HN-1 (bis(2-choroethyl)ethylamine), HN-2 and HN-3 undergo hydrolysis to form