Merck
14505
Sigma-Aldrich
Sodium stannate
42-45% SnO2 basis
About This Item
Fórmula lineal:
Na2SnO3
Número de CAS:
Peso molecular:
212.69
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22
Quality Level
form
powder
composition
SnO2, 42-45%
reaction suitability
core: tin
reagent type: catalyst
storage temp.
2-8°C
SMILES string
[Na+].[Na+].[O-][Sn]([O-])=O
InChI
1S/2Na.3O.Sn/q2*+1;;2*-1;
InChI key
TVQLLNFANZSCGY-UHFFFAOYSA-N
pictograms
CorrosionEnvironment
signalword
Danger
hcodes
Hazard Classifications
Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1
Storage Class
8B - Non-combustible corrosive hazardous materials
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Kejun Chen et al.
Nature communications, 11(1), 4173-4173 (2020-08-21)
Iron phthalocyanine (FePc) is a promising non-precious catalyst for the oxygen reduction reaction (ORR). Unfortunately, FePc with plane-symmetric FeN4 site usually exhibits an unsatisfactory ORR activity due to its poor O2 adsorption and activation. Here, we report an axial Fe-O
Lu Liu et al.
Polymers, 11(2) (2019-04-10)
Manganese dioxide (MnO₂), as a promising green material, has recently attracted considerable attention of researchers from various fields. In this work, a facile method was introduced to prepare binary hybrids by fabricating three-dimensional (3D) zinc hydroxystannate (ZHS) cubes on two-dimensional
Wenjin Wan et al.
Nanomaterials (Basel, Switzerland), 8(2) (2018-02-22)
Two dimensional (2D)SnO2 nanosheets were synthesized by a substrate-free hydrothermal route using sodium stannate and sodium hydroxide in a mixed solvent of absolute ethanol and deionized water at a lower temperature of 130 °C. The characterization results of the morphology
Wei Wang et al.
Journal of hazardous materials, 340, 263-271 (2017-07-18)
Zinc hydroxystannate (ZHS) was fabricated on the surface of amorphous hydrous TiO
Min-Fu Wu et al.
Analytica chimica acta, 1049, 188-195 (2019-01-08)
An electrochemical immunosensor for ultrasensitive detection of acrylamide (AA) in water and food samples was developed. SnO2-SiC hollow sphere nanochains with high surface area and gold nanoparticles with good electroconductivity were fabricated onto the surface of a glassy carbon electrode