Merck
MDE3007FS1
Millipore
Filtro de profundidad Millistak+® Pod
DE30, filtration area 0.77m², Process Scale
About This Item
Código UNSPSC:
23151806
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Nombre del producto
Filtro de profundidad Millistak+® Pod, DE30 (media series), cellulose membrane (and inorganic filter aid), effective filtration area 0.77 m2
Materiales
TPE (Thermo Plastic Elastomer)
TPE gasket (Thermo Plastic Elastomer)
cellulose membrane (and inorganic filter aid)
polypropylene housing (glass-filled)
Nivel de calidad
Compatibilidad de esterilización
autoclavable compatible
área de filtración efectiva
0.77 m2
Adaptador
seal (flat seal)
código del cartucho
DE30 (media series)
Descripción general
The Millistak+® depth filter media is offered in a scalable, disposable format providing greater flexibility because of its unique modular design. The Millistak+® MDE3007FS1 comes in a POD format.
This DE30 media series delivers optimal performance through a gradient density matri, ideal for primary or secondary clarification applications. Millistak+® Pod filters are available in three distinct series of media grades in order to meet your specific needs - DE, CE and HC.
The Millistak+® Pod system is ideal for a wide variety of primary and secondary clarification applications, including cell cultures, yeast and E. coli lysates post centrifuge, E. coli refolds, media, vaccines, plasma proteins and sera.
Easy to Use
With the compact, modular design of our Millistak+® new Pod system, you can increase productivity and shorten cycle times.
Installation and setup of the Pod system is simple and straightforward. The unique design of the disposable adapters and disposable manifolds makes it easy to connect the Pod filters to rest of the unit operations in the process. The self-contained and disposable nature of the system protects operators from exposure to biohazards and eliminates maintenance as well as cleaning validation requirements.
Características y beneficios
  • Low hold-up volume for greater product yield
  • Broad range of media types offered in single and multilayer products
  • Flexible, modular format offers scalability from 5 to 12,000 liters or more
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Self-contained Pod filters protect operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces
Información legal
MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany
¿Qué es el programa Emprove®?
El programa Emprove® es un sistema que proporciona documentación completa y exhaustiva de nuestros filtros y componentes de un solo uso, materias primas farmacéuticas y materiales de partida.
Material Qualification Dossier
Gratuito, iniciar sesión para descargar
Más información sobre las ventajas que le puede proporcionar una suscripción al programa Emprove®
¿Por qué no están disponibles estos documentos?
Los expedientes Emprove® contienen información destinada a respaldar el proceso de registro y las actividades de evaluación del riesgo de los productores farmacéuticos y contienen información confidencial. contacte con nosotros si desea tener acceso.