Documentos clave
Z654892
Best® Nitri-Care® gloves
size 7‑8 M
About This Item
Código UNSPSC:
46181504
Materiales
nitrile
tamaño
7‑8 M
esterilidad
non-sterile
Características
CE compliant
disposable
latex free
powder-free
envase
package of 100 ea
fabricante / nombre comercial
Best Manufacturing 3005PFM
L × grosor
9.5 in. × 4 mil
color
blue
Descripción general
  • Tested according to EN 455
  • Ambidextrous
  • The "low-modulus" feature: the glove becomes a second skin
  • Nitri-Care provides three times the puncture resistance of natural rubber and vinyl gloves of comparable thickness (puncture resistance is > 0.45 Kg)
  • Best protection against latex allergies, viruses and bacteria
Nitri-Care is the economical medical examination version of the N-DEX family. Made in ISO 9002 quality-certified facility, Nitri-Care is inspected to a quality level of 1.5AQL. Nitri-Care does not contain any proteins of natural rubber and features the proprietary non-allergenic polymeric coating that aids in wet-hand donning.
  • Tested according to EN 455
  • Ambidextrous
  • The "low-modulus" feature: the glove becomes a second skin
  • Nitri-Care provides three times the puncture resistance of natural rubber and vinyl gloves of comparable thickness (puncture resistance is > 0.45 Kg)
  • Best protection against latex allergies, viruses and bacteria
Información legal
Best is a registered trademark of Showa Glove Co.
Nitri-Care is a registered trademark of Best Manufacturing Co.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)