Documentos
Z189987
ACE heavy-wall powder funnels
Mini-Lab, top diam. 60 mm
About This Item
UNSPSC Code:
41121809
NACRES:
NB.43
joint
joint ST/NS14/20
manufacturer/tradename
Ace Glass 948810
top diam.
60 mm
General description
For pouring powders or liquids into ground-joint containers.