Documentos clave
Z155950
Stainless steel drying trays
rolled bead, L × W × H 13.7 in. × 9.2 in. × 0.75 in.
About This Item
Código UNSPSC:
41104510
NACRES:
NB.43
En este momento no podemos mostrarle ni los precios ni la disponibilidad
fabricante / nombre comercial
Polarware 13F
L × An × Al
13.7 in. × 9.2 in. × 0.75 in.
Descripción general
Stainless steel drying trays have a low profile, and are lighter weight than glass. We carry a wide range of sizes. The unbreakable stainless steel material has rapid cooling and heating for your samples.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)