Z105635
Disposable apron
polyethylene, pkg of 100 ea
About This Item
Código UNSPSC:
41100000
Productos recomendados
Materiales
polyethylene
envase
pkg of 100 ea
disponibilidad
not available in EU, EU
Descripción general
One-piece, liquid-proof PE apron resists many chemicals, fats, greases, and oils. Extra-long ties, embossed finish. Packaged in convenient dispenser of 100 aprons.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)