Documentos clave
TEM-LC400-NI-100
Sigma-Aldrich
Lacey Carbon Supported Nickel Grid
size 400 mesh, hole size 100 μm , Average, box of 25
Seleccione un Tamaño
1 PKG
MXP 1,382.00
MXP 1,382.00
Check Cart for Availability
Solicitar un pedido a granel
Seleccione un Tamaño
Cambiar Vistas
1 PKG
MXP 1,382.00
About This Item
Código UNSPSC:
41111761
NACRES:
MA.24
MXP 1,382.00
Check Cart for Availability
Solicitar un pedido a granel
envase
box of 25
agujero tamaño
100 μm , Average
tamaño
400 mesh
Descripción general
Lacey Carbon Supported Grid: Nickel Grid, 400 mesh. Average hole size is 100 micron. This carbon coated film on a broken pattern consists of woven-mesh-like openings of different sizes and shapes. This type of pattern provides support but does not interfere when observing specimen sections.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)