Documentos clave
TEM-G300F1-NI
Sigma-Aldrich
Gilder Finder Grids G300F1
Nickel, size 300 mesh, vial of 100
About This Item
UNSPSC Code:
41111761
packaging
vial of 100
size
300 mesh
General description
Gilder Finder Grids G300F1. Nickel grids, 300 mesh. The block of 625 (25x25) individual cells is divided into smaller blocks bounded by thicker crenellated bars. Each small block of 25 cells has a central alphabet letter from A to Y for identification purposes.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)