Merck
T299308
2,3-DIPHENYL-2-CYCLOPROPEN-1-YLIDENESULFAMOYL CHLORIDE
AldrichCPR
Seleccione un Tamaño
5MG
MXP 1,646.00
MXP 1,646.00
Fecha estimada de envío05 de abril de 2025
Seleccione un Tamaño
Cambiar Vistas
5MG
MXP 1,646.00
About This Item
Fórmula lineal:
C15H10ClNO2S
Número de CAS:
Peso molecular:
303.769
MDL number:
MXP 1,646.00
Fecha estimada de envío05 de abril de 2025
Other Notes
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)