Merck
T147192
1-(4-PYRIDINYL)ETHANONE [4-(4-CHLOROPHENYL)-1,3-THIAZOL-2-YL]HYDRAZONE
AldrichCPR
Seleccione un Tamaño
10MG
MXP 681.00
MXP 681.00
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
10MG
MXP 681.00
About This Item
Fórmula lineal:
C16H13ClN4S
Número de CAS:
Peso molecular:
328.826
Número MDL:
MXP 681.00
Check Cart for Availability
Otras notas
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)