Merck
T120499
2-CHLORO-N-(4-((2-CHLOROBENZOYL)AMINO)-3-NITROPHENYL)BENZAMIDE
AldrichCPR
Seleccione un Tamaño
50MG
MXP 1,646.00
MXP 1,646.00
Fecha estimada de envío22 de mayo de 2025
Seleccione un Tamaño
Cambiar Vistas
50MG
MXP 1,646.00
About This Item
Fórmula lineal:
C20H13Cl2N3O4
Número de CAS:
Peso molecular:
430.25
Número MDL:
MXP 1,646.00
Fecha estimada de envío22 de mayo de 2025
Otras notas
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)