Documentos clave
CLS606090
Pyrex® Buchner funnel, with perforated plate
About This Item
Código UNSPSC:
41121809
NACRES:
NB.43
fabricante / nombre comercial
Corning 6060-90
Diámetro
90.0 mm
color
clear
Descripción general
With perforated plate. Suitable for vacuum filtration, but translucent plate should not be subjected to pressure differential in excess of 1atm.
Información legal
PYREX is a registered trademark of Corning, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)