Documentos clave
CAD4182
Stainless steel 304 syringe needle
gauge 20, L 1 in.
Seleccione un Tamaño
12 EA
MXP 6,234.00
MXP 6,234.00
Fecha estimada de envío18 de abril de 2025
Seleccione un Tamaño
Cambiar Vistas
12 EA
MXP 6,234.00
About This Item
UNSPSC Code:
42142600
NACRES:
NB.43
MXP 6,234.00
Fecha estimada de envío18 de abril de 2025
description
Standard Hub with 304 Stainless Steel Tubing
packaging
pack of 12 ea
manufacturer/tradename
Cadence Science 4182
L
1 in.
gauge
20
General description
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved, deflecting tip to prevent coring during needle insertion through the septum.
Standard Hub with 304 Stainless Steel Tubing
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)