Documentos clave
BR13956
BRAND® spare burette tubes for compact burette
size 25 mL, SILBERBRAND, amber AR-Glass
Sinónimos:
Accessories, Burettes, Compact Burettes, SILBERBRAND
Seleccione un Tamaño
1 EA
MXP 1,509.00
MXP 1,509.00
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
1 EA
MXP 1,509.00
About This Item
Código UNSPSC:
41121808
NACRES:
NB.43
MXP 1,509.00
Check Cart for Availability
Materiales
amber AR-Glass
envase
pack of 1 ea
fabricante / nombre comercial
BRAND 13956
Longitud
425 mm
tamaño
25 mL
Categorías relacionadas
Descripción general
The BRAND® spare burette tubes for compact burettes are crafted from high-quality Boro 3.3 amber glass, ensuring both durability and visibility for accurate liquid measurement. These spare tubes are specifically designed to be compatible with BLAUBRAND® and SILBERBRAND compact burettes, making them a reliable choice for maintaining your liquid handling systems.
Información legal
BRAND is a registered trademark of BRAND GMBH + CO KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)