BAF183070003
Scienceware® Vikem® coated closed lead ring
for fits flask 5-10 mL
About This Item
Código UNSPSC:
41122800
NACRES:
NB.43
Productos recomendados
envase
pack of 1 ea
fabricante / nombre comercial
Bel-Art F18307-0003
Adaptador
for fits flask 5-10 mL
Descripción general
Protect and stabilize expensive glassware with these Vikem® vinyl coated lead rings.
Características y beneficios
- Stabilize glassware immersed in water baths;reduces floating, breakage and chemical spills
- Bright red Vikem® vinyl coating
- Chemical-resistant
- Will not scratch flasks
Información legal
Scienceware is a registered trademark of SP Scienceware, a division of SP
Vikem is a registered trademark of SP Scienceware, a division of SP
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)