Merck
328847
Sigma-Aldrich
(S)-(−)-Pulegone
98%
Sinónimos:
(−)-Pulegone, (S)-2-Isopropylidene-5-methylcyclohexanone, (S)-p-Menth-4(8)-en-3-one
About This Item
Fórmula empírica (notación de Hill):
C10H16O
Número de CAS:
Peso molecular:
152.23
Beilstein/REAXYS Number:
2040703
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.22
assay
98%
optical activity
[α]20/D −22°, neat
refractive index
n20/D 1.488 (lit.)
bp
223-224 °C (lit.)
density
0.937 g/mL at 25 °C (lit.)
SMILES string
C[C@H]1CC\C(=C(/C)C)C(=O)C1
InChI
1S/C10H16O/c1-7(2)9-5-4-8(3)6-10(9)11/h8H,4-6H2,1-3H3/t8-/m0/s1
InChI key
NZGWDASTMWDZIW-QMMMGPOBSA-N
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Visite la Librería de documentos
Yi Wang et al.
Plant, cell & environment, 44(2), 559-573 (2020-11-21)
In plants, cellular lipid peroxidation is enhanced under low nitrogen (LN) stress; this increases the lipid-derived reactive carbonyl species (RCS) levels. The cellular toxicity of RCS can be reduced by various RCS-scavenging enzymes. However, the roles of these enzymes in
Mitscheli S Da Rocha et al.
Toxicological sciences : an official journal of the Society of Toxicology, 128(1), 1-8 (2012-04-14)
Essential oils from mint plants, including peppermint and pennyroyal oils, are used at low levels as flavoring agents in various foods and beverages. Pulegone is a component of these oils. In a 2-year bioassay, oral administration of pulegone slightly increased
Chang-Qin Li et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 37(21), 3173-3175 (2013-02-13)
To study the validity of Schizonepeta tenuifalia pieces. The change principle for the contents of essential oil and pulegone was determined by the way of classical constant temperature acceleration experiment, and the reacting speed at 20 degrees C was calculated
Barbara Kay Clapperton et al.
Pest management science, 68(6), 870-877 (2012-02-18)
Repellents may prevent bird pests from eating crops or protect non-target birds from eating harmful substances. The feeding behaviour of free-ranging house sparrows (Passer domesticus) presented with wheat treated with the secondary repellent anthraquinone (AQ), paired with visual and/or olfactory
Xiaoying Zhou et al.
Natural product communications, 7(1), 81-82 (2012-03-21)
The essential oils of Ziziphora clinopodioide Lam. from four different production areas (Banfang ditch; Altay mountains; Tuoli; Terks) were investigated. The oils were extracted by hydro-distillation and analyzed by gas chromatography-mass spectrometry (GC-MS). Seventeen constituents were identified in the essential