Skip to Content
Merck
Bis(cyclopentadienyl)titanium(IV) bis(trifluoromethanesulfonate) 98%

Bis(cyclopentadienyl)titanium(IV) bis(trifluoromethanesulfonate)

Bis(cyclopentadienyl)titanium(IV) bis(trifluoromethanesulfonate) 98%
Synonym(s):
Bis(cyclopentadienyl)di(triflato)titanium, Di(cyclopentadienyl)titanium(IV) bis(trifluoromethanesulfonate), Titanocene bis(trifluoromethanesulfonate), Titanocene ditriflate
Empirical Formula (Hill Notation):
C12H10F6O6S2Ti
CAS Number:
Molecular Weight:
476.19

Products

Brand
Formula Weight
Purity
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing