Skip to Content
Merck
All Photos(1)

Key Documents

V000470

Sigma-Aldrich

Xylenol Orange tetrasodium salt

p.a., ACS reagent

Synonym(s):

3,3′-Bis[N,N-bis(carboxymethyl)aminomethyl]-o-cresolsulfonephthalein tetrasodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H28N2Na4O13S
CAS Number:
Molecular Weight:
760.58
EC Number:
MDL number:
PubChem Substance ID:

Pricing and availability is not currently available.

grade

ACS reagent
p.a.

mp

195 °C (dec.) (lit.)

SMILES string

[Na+].[Na+].[Na+].[Na+].Cc1cc(cc(CN(CC([O-])=O)CC([O-])=O)c1O)C2(OS(=O)(=O)c3ccccc23)c4cc(C)c(O)c(CN(CC([O-])=O)CC([O-])=O)c4

InChI

1S/C31H32N2O13S.4Na/c1-17-7-21(9-19(29(17)42)11-32(13-25(34)35)14-26(36)37)31(23-5-3-4-6-24(23)47(44,45)46-31)22-8-18(2)30(43)20(10-22)12-33(15-27(38)39)16-28(40)41;;;;/h3-10,42-43H,11-16H2,1-2H3,(H,34,35)(H,36,37)(H,38,39)(H,40,41);;;;/q;4*+1/p-4

InChI key

YIIQUYHVWRJXEM-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service