1394008
USP
Mestranol
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
(17α)-3-Methoxy-19-norpregna-1,3,5(10)-trien-20-yn-17-ol, 17α-Ethynyl-1,3,5(10)-estratriene-3,17β-diol 3-methyl ether, 17α-Ethynylestradiol 3-methyl ether, 3-Methoxy-17α-ethynylestradiol, NSC 84032
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
mestranol
manufacturer/tradename
USP
mp
153-155 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
SMILES string
COc1ccc2C3CC[C@@]4(C)C(CC[C@@]4(O)C#C)C3CCc2c1
InChI
1S/C21H26O2/c1-4-21(22)12-10-19-18-7-5-14-13-15(23-3)6-8-16(14)17(18)9-11-20(19,21)2/h1,6,8,13,17-19,22H,5,7,9-12H2,2-3H3/t17-,18-,19+,20+,21+/m1/s1
InChI key
IMSSROKUHAOUJS-MJCUULBUSA-N
Gene Information
human ... ESR1(2099)
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
- Norethindrone and Mestranol Tablets
Analysis Note
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service